Alfa Aesar™ 2,6-Di-tert-butylpyridine, 97+%

Manufacturer: Alfa Aesar™ H5576106

Quantity 5g

 View more versions of this product

Catalog No. AAH5576106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 585-48-8
    Molecular Formula C13H21N
    Formula Weight 191.32
    MDL Number MFCD00006306
    Beilstein 125886
    Merck Index 14,3036
    Chemical Name or Material 2,6-Di-tert-butylpyridine

    Specifications

    Formula Weight 191.32
    Merck Index 14,3036
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Flash Point 71°C (160°F)
    Molecular Formula C13H21N
    MDL Number MFCD00006306
    CAS 585-48-8
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 100° to 101°C (23mmHg)
    Quantity 5g
    Beilstein 125886
    Chemical Name or Material 2,6-Di-tert-butylpyridine
    Assay ≥97%

    Description

    Applications
    2,6-Di-tert-butylpyridine is used in the preparation of 2, 6-di-tert-butylpyridine hydrotriflate. It is used as a proton scavenger to check the progress of the living polymerization of isobutylene. It is associated with cerric ammonium nitrate and used in the alfa-enolation of aldehydes. It is involved in the preparation of vinyl triflate using polymer-bound 2,6-di-tert-butylpyridine.

    Solubility
    Miscible with alcohol, acetone, and hexane. Immiscible with water.

    Notes
    Store in a cool place. Incompatible with strong acids and strong oxidizing agents.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit