Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Di-tert-butylpyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
585-48-8
C13H21N
191.32
MFCD00006306
UWKQJZCTQGMHKD-UHFFFAOYSA-N
2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119
68510
2,6-ditert-butylpyridine
CC(C)(C)C1=NC(=CC=C1)C(C)(C)C
585-48-8
C13H21N
191.32
MFCD00006306
UWKQJZCTQGMHKD-UHFFFAOYSA-N
2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119
68510
2,6-ditert-butylpyridine
CC(C)(C)C1=NC(=CC=C1)C(C)(C)C
Specific Gravity 0.852
Packaging Glass bottle
Quantity 5g
Melting Point 2.0°C to 4.0°C
Flash Point 72°C
Refractive Index 1.4720 to 1.4740
Color Yellow
Infrared Spectrum Authentic
Boiling Point 208.0°C
Density 0.8520g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (C5H3N[C(CH3)3]2
Beilstein 20, III, 2868
Fieser 01,212
Merck Index 15, 3045
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Formula Weight 191.32
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2, 6-Di-tert-butylpyridine, 97%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 209-557-6