Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Diamino-3,5-difluoropyridine, 98%

Manufacturer:  Alfa Aesar™ H3036503

Quantity 1g

 View more versions of this product

Catalog No. AAH3036503


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

247069-27-8
145.11
UN2811
2,6-Diamino-3,5-difluoropyridine
C5H5F2N3
MFCD03412208
3,5-Difluoro-2,6-pyridinediamine

Specifications

158° to 162°C
247069-27-8
145.11
UN2811
2,6-Diamino-3,5-difluoropyridine
1g
C5H5F2N3
MFCD03412208
3,5-Difluoro-2,6-pyridinediamine
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit