Alfa Aesar™ 2,6-Diaminopyridine, 98%

Manufacturer: Alfa Aesar™ A1229536

Quantity 500g

 View more versions of this product

Catalog No. AAA1229536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 141-86-6
    Molecular Formula C5H7N3
    Formula Weight 109.13
    MDL Number MFCD00006329
    UN Number UN2671
    Beilstein 108513
    Synonym 2,6-Pyridinediamine
    Chemical Name or Material 2,6-Diaminopyridine

    Specifications

    Formula Weight 109.13
    Health Hazard 3 GHS P Statement
    P260-P261-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 141-86-6
    Molecular Formula C5H7N3
    MDL Number MFCD00006329
    Synonym 2,6-Pyridinediamine
    UN Number UN2671
    Beilstein 108513
    Melting Point 118° to 121°C
    TSCA TSCA
    Odor Unpleasant
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H373-H319-H335
    Toxic if swallowed.
    May cause damage to organs through prolonged or repeated exposure.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES
    Flash Point 155°C (311°F)
    RTECS Number US7570000
    Boiling Point 288°C
    Quantity 500g
    Chemical Name or Material 2,6-Diaminopyridine
    Assay 98%

    Description

    Applications
    2,6-Diaminopyridine, used as a pharmaceutical intermediate and a hair dye coupler in oxidation/permanent formulations.

    Solubility
    Soluble in water 180 g/L @ 20°C, acetone, ethanol, methanol, isopropanol, ethyl acetate.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

    GHS H Statement
    H301-H373-H319-H335
    Toxic if swallowed.
    May cause damage to organs through prolonged or repeated exposure.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P261-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit