Alfa Aesar™ 2',6-Dibromo-3,4'-bipyridine, 95%

Manufacturer: Alfa Aesar™ H27214MC

Quantity 100mg

 View more versions of this product

Catalog No. AAH27214MC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 942206-16-8
    Molecular Formula C10H6Br2N2
    Formula Weight 313.98
    MDL Number MFCD09037424
    Chemical Name or Material 2',6-Dibromo-3,4'-bipyridine

    Specifications

    Formula Weight 313.98
    Melting Point 189° to 193°C
    Quantity 100mg
    Molecular Formula C10H6Br2N2
    MDL Number MFCD09037424
    CAS 942206-16-8
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2',6-Dibromo-3,4'-bipyridine
    Assay 95%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit