Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dibromo-4-fluorophenol, 98+%

Click to view available options
Quantity:
5g
25g
100g
344-20-7
C6H3Br2FO
269.895
MFCD00002317
RRAZCUUOWIDAJS-UHFFFAOYSA-N
phenol, 2,6-dibromo-4-fluoro,2,6-dibromo-4-fluoro-phenol,pubchem2868,acmc-209i7j,ksc494q8h,emolecules 492569,1-bromo-3,4,5-trifluorobezene,attercop-chm at113643,2,6-dibromo-4-fluorophenol
67654
2,6-dibromo-4-fluorophenol
C1=C(C=C(C(=C1Br)O)Br)F
344-20-7
C6H3Br2FO
269.895
MFCD00002317
RRAZCUUOWIDAJS-UHFFFAOYSA-N
phenol, 2,6-dibromo-4-fluoro,2,6-dibromo-4-fluoro-phenol,pubchem2868,acmc-209i7j,ksc494q8h,emolecules 492569,1-bromo-3,4,5-trifluorobezene,attercop-chm at113643,2,6-dibromo-4-fluorophenol
67654
2,6-dibromo-4-fluorophenol
C1=C(C=C(C(=C1Br)O)Br)F
Quantity 5g
Odor Phenol-like
Melting Point 54°C to 56°C
Beilstein 1868033
Formula Weight 269.91
Percent Purity ≥98%
Chemical Name or Material 2,6-Dibromo-4-fluorophenol

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger