Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dibromo-4-methylphenol, 98%

Click to view available options
Quantity:
25g
100g
2432-14-6
C7H6Br2O
265.932
MFCD00002154
FIGPGTJKHFAYRK-UHFFFAOYSA-N
2,6-dibromo-p-cresol,dibromocresol,p-cresol, 2,6-dibromo,3,5-dibromo-4-hydroxytoluene,phenol, 2,6-dibromo-4-methyl,2,6-dibromo-4-methyl-phenol,attercop-chm at113597,dibromopcresol,p-cresol,6-dibromo,2,6-dibromo-p-creso
17078
2,6-dibromo-4-methylphenol
CC1=CC(=C(C(=C1)Br)O)Br
2432-14-6
C7H6Br2O
265.932
MFCD00002154
FIGPGTJKHFAYRK-UHFFFAOYSA-N
2,6-dibromo-p-cresol,dibromocresol,p-cresol, 2,6-dibromo,3,5-dibromo-4-hydroxytoluene,phenol, 2,6-dibromo-4-methyl,2,6-dibromo-4-methyl-phenol,attercop-chm at113597,dibromopcresol,p-cresol,6-dibromo,2,6-dibromo-p-creso
17078
2,6-dibromo-4-methylphenol
CC1=CC(=C(C(=C1)Br)O)Br
Flash Point >110°C (230°F)
Melting Point 49°C to 50°C
Quantity 100g
Beilstein 2045283
Formula Weight 265.94
Percent Purity 98%
Chemical Name or Material 2,6-Dibromo-4-methylphenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 219-404-5

RTECSNumber : GO7700000

TSCA : No

Recommended Storage : Ambient temperatures