Alfa Aesar™ 2,6-Dibromo-4-n-propylaniline, 98%

Manufacturer: Alfa Aesar™ B2480214

Quantity 25g

 View more versions of this product

Catalog No. AAB2480214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10546-64-2
    Molecular Formula C9H11Br2N
    Formula Weight 293.01
    MDL Number MFCD00051706
    Chemical Name or Material 2,6-Dibromo-4-n-propylaniline

    Specifications

    Formula Weight 293.01
    Melting Point 55° to 57°C
    MDL Number MFCD00051706
    CAS 10546-64-2
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 25g
    Molecular Formula C9H11Br2N
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2,6-Dibromo-4-n-propylaniline
    Assay 98%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit