Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dibromophenol, 99%

Click to view available options
Quantity:
1g
5g
25g
608-33-3
C6H4Br2O
251.905
MFCD00002152
SSIZLKDLDKIHEV-UHFFFAOYSA-N
phenol, 2,6-dibromo,2,6-dibromo-phenol,unii-27mip05eav,2,6-dibromo-pheno,27mip05eav,2,6-bis bromanyl phenol,acmc-1au9i,2,6-dibromophenol,ksc493e5t,2,6-dibromophenol, analytical standard
11847
CHEBI:19391
2,6-dibromophenol
C1=CC(=C(C(=C1)Br)O)Br
608-33-3
C6H4Br2O
251.905
MFCD00002152
SSIZLKDLDKIHEV-UHFFFAOYSA-N
phenol, 2,6-dibromo,2,6-dibromo-phenol,unii-27mip05eav,2,6-dibromo-pheno,27mip05eav,2,6-bis bromanyl phenol,acmc-1au9i,2,6-dibromophenol,ksc493e5t,2,6-dibromophenol, analytical standard
11847
CHEBI:19391
2,6-dibromophenol
C1=CC(=C(C(=C1)Br)O)Br
Applications
2,6-Dibromophenol is used in the production of 3,4,5-trimethoxybenzaldehyde and 2-(2,6-dibromophenoxy)ethylbromide.

Solubility
Soluble in ethanol and ether. Slightly soluble in water.

Notes
Incompatible with acid chlorides, acid anhydrides and oxidizing agents.
Odor Phenol-like
Boiling Point 255°C to 256°C
Melting Point 53°C to 57°C
Quantity 1g
Beilstein 2043614
Solubility Information Soluble in ethanol and ether. Slightly soluble in water.
Formula Weight 251.92
Percent Purity 99%
Chemical Name or Material 2,6-Dibromophenol

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 210-161-0

TSCA : Yes

Recommended Storage : Ambient temperatures