Alfa Aesar™ 2,6-Dibromotoluene, 98+%

Manufacturer: Alfa Aesar™ A1183203

Quantity 1g

 View more versions of this product

Catalog No. AAA1183203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 69321-60-4
    Molecular Formula C7H6Br2
    Formula Weight 249.94
    MDL Number MFCD00013524
    Beilstein 3235502
    Chemical Name or Material 2,6-Dibromotoluene

    Specifications

    Formula Weight 249.94
    CAS 69321-60-4
    Boiling Point 112° to 113°C (7mmHg)
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    MDL Number MFCD00013524
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Molecular Formula C7H6Br2
    Beilstein 3235502
    Melting Point 2° to 6°C
    Health Hazard 1 Warning
    Chemical Name or Material 2,6-Dibromotoluene
    Assay ≥98%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit