Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dichloro-3-cyanopyridine, 97%

Click to view available options
Quantity:
250mg
1g
5g
40381-90-6
C6H2Cl2N2
172.996
MFCD03411341
LFCADBSUDWERJT-UHFFFAOYSA-N
2,6-dichloronicotinonitrile,2,6-dichloro-3-cyanopyridine,3-pyridinecarbonitrile, 2,6-dichloro,2,6-dichloro-nicotinonitrile,2,6-dichloro-3-pyridinecarbonitrile,2,6-dichloropyridine-3-carbonitrile,pubchem14370,acmc-20a0s7,ksc495i4d,3-cyano-2,6-dichloropyridine
12605538
2,6-dichloropyridine-3-carbonitrile
C1=CC(=NC(=C1C#N)Cl)Cl
40381-90-6
C6H2Cl2N2
172.996
MFCD03411341
LFCADBSUDWERJT-UHFFFAOYSA-N
2,6-dichloronicotinonitrile,2,6-dichloro-3-cyanopyridine,3-pyridinecarbonitrile, 2,6-dichloro,2,6-dichloro-nicotinonitrile,2,6-dichloro-3-pyridinecarbonitrile,2,6-dichloropyridine-3-carbonitrile,pubchem14370,acmc-20a0s7,ksc495i4d,3-cyano-2,6-dichloropyridine
12605538
2,6-dichloropyridine-3-carbonitrile
C1=CC(=NC(=C1C#N)Cl)Cl
Sensitivity Moisture sensitive
Quantity 250mg
Melting Point 121°C to 125°C
UN Number UN3439
Formula Weight 173
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-3-cyanopyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon