Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-3-fluoro-5-(trifluoromethyl)pyridine, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
1099598-11-4
C6HCl2F4N
233.98
VVXQCTFINCODIF-UHFFFAOYSA-N
2,6-dichloro-3-fluoro-5-trifluoromethyl pyridine
37859559
2,6-dichloro-3-fluoro-5-(trifluoromethyl)pyridine
C1=C(C(=NC(=C1F)Cl)Cl)C(F)(F)F
1099598-11-4
C6HCl2F4N
233.98
VVXQCTFINCODIF-UHFFFAOYSA-N
2,6-dichloro-3-fluoro-5-trifluoromethyl pyridine
37859559
2,6-dichloro-3-fluoro-5-(trifluoromethyl)pyridine
C1=C(C(=NC(=C1F)Cl)Cl)C(F)(F)F
Boiling Point 90°C (55.0 mbar)
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 233.98
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloro-3-fluoro-5-(trifluoromethyl)pyridine

GHS Signal Word: Warning