Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-3-fluoroacetophenone, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
290835-85-7
C8H5Cl2FO
207.03
MFCD02093760
VJBFZHHRVCPAPZ-UHFFFAOYSA-N
2',6'-dichloro-3'-fluoroacetophenone,1-2,6-dichloro-3-fluorophenyl ethanone,2,6-dichloro-3-fluoroacetophenone,1-2,6-dichloro-3-fluorophenyl ethan-1-one,2,6-dichloro-3-fluro acetophenone,2,6-dichloro-5-fluoro acetophenone,2',6'-dichloro-3'-fluroacetophenone,ethanone, 1-2,6-dichloro-3-fluorophenyl,1-acetyl-2,6-dichloro-3-fluorobenzene,pubchem8532
2733982
1-(2,6-dichloro-3-fluorophenyl)ethanone
CC(=O)C1=C(C=CC(=C1Cl)F)Cl
290835-85-7
C8H5Cl2FO
207.03
MFCD02093760
VJBFZHHRVCPAPZ-UHFFFAOYSA-N
2',6'-dichloro-3'-fluoroacetophenone,1-2,6-dichloro-3-fluorophenyl ethanone,2,6-dichloro-3-fluoroacetophenone,1-2,6-dichloro-3-fluorophenyl ethan-1-one,2,6-dichloro-3-fluro acetophenone,2,6-dichloro-5-fluoro acetophenone,2',6'-dichloro-3'-fluroacetophenone,ethanone, 1-2,6-dichloro-3-fluorophenyl,1-acetyl-2,6-dichloro-3-fluorobenzene,pubchem8532
2733982
1-(2,6-dichloro-3-fluorophenyl)ethanone
CC(=O)C1=C(C=CC(=C1Cl)F)Cl
Packaging Glass bottle
Refractive Index 1.5275 to 1.5295
Boiling Point 250.0°C to 255.0°C
Quantity 100g
Flash Point 110°C
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 207.03
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloro-3-fluoroacetophenone, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA