Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dichloro-3-iodopyridine, 98%

Click to view available options
Quantity:
1g
5g
148493-37-2
C5H2Cl2IN
273.882
MFCD03094686
DPCQIHCGMIPSQV-UHFFFAOYSA-N
2,6-dichloro-3-iodo-pyridine,pyridine, 2,6-dichloro-3-iodo,zlchem 993,acmc-20aihk,pubchem14239,tpc-py080,2,6-dichloro-3-iodo pyridine,pyridine,2,6-dichloro-3-iodo,2,6-dichloro-3-iodopyridine
2736002
2,6-dichloro-3-iodopyridine
C1=CC(=NC(=C1I)Cl)Cl
148493-37-2
C5H2Cl2IN
273.882
MFCD03094686
DPCQIHCGMIPSQV-UHFFFAOYSA-N
2,6-dichloro-3-iodo-pyridine,pyridine, 2,6-dichloro-3-iodo,zlchem 993,acmc-20aihk,pubchem14239,tpc-py080,2,6-dichloro-3-iodo pyridine,pyridine,2,6-dichloro-3-iodo,2,6-dichloro-3-iodopyridine
2736002
2,6-dichloro-3-iodopyridine
C1=CC(=NC(=C1I)Cl)Cl
Applications
It is used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Quantity 1g
Color White to Brown
Melting Point 74°C to 79°C
Sensitivity Light Sensitive
UN Number UN2811
Beilstein 6135696
Formula Weight 273.88
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-3-iodopyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C