Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-3-nitropyridine, 90%, Tech., ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
16013-85-7
C5H2Cl2N2O2
192.99
MFCD00006234
SHCWQWRTKPNTEM-UHFFFAOYSA-N
2,6-dichloro-3-nitro-pyridine,pyridine, 2,6-dichloro-3-nitro,3-nitro-2,6-dichloropyridine,2,6-dichloro-3-nitro pyridine,zlchem 223,pubchem9234,acmc-209dko,2,6-dichloronitropyridine,dichloronitropyridine,ksc174q3n
85239
2,6-dichloro-3-nitropyridine
C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl
16013-85-7
C5H2Cl2N2O2
192.99
MFCD00006234
SHCWQWRTKPNTEM-UHFFFAOYSA-N
2,6-dichloro-3-nitro-pyridine,pyridine, 2,6-dichloro-3-nitro,3-nitro-2,6-dichloropyridine,2,6-dichloro-3-nitro pyridine,zlchem 223,pubchem9234,acmc-209dko,2,6-dichloronitropyridine,dichloronitropyridine,ksc174q3n
85239
2,6-dichloro-3-nitropyridine
C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl
Flash Point >110°C
Infrared Spectrum Authentic
Melting Point 58°C to 68°C
Quantity 250g
Water 10% Max. (K.F.)
Packaging Glass bottle
Color Colorless to Yellow
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Linear Formula Cl2(C5H2N)NO2
Formula Weight 192.99
Physical Form Liquid
Percent Purity 90%
Grade Technical
Chemical Name or Material 2, 6-Dichloro-3-nitropyridine
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position com

GHS Signal Word: Warning

EINECSNumber : 240-151-1