Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dichloro-3-(trifluoromethyl)benzyl bromide, 97%

Manufacturer:  Alfa Aesar™ H3179206

Quantity 5g

 View more versions of this product

Catalog No. AAH3179206


  Check Availability
Add to cart

Specifications

Description

Applications
2,6-Dichlorobenzyl Bromide is used in the preparation of potent epoxide hydrolyses inhibitors. Has shown thus far to relieve hypotension in lipopolysaccharide deficient murine models. And it is used in the synthesis of anti-HIV drugs based on diaryltriazine (DATA) analogues.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

C8H4BrCl2F3
MFCD11519349
2,6-Dichloro-3-(trifluoromethyl)benzyl bromide
307.93
UN3261

Specifications

5g
307.93
UN3261
2,6-Dichloro-3-(trifluoromethyl)benzyl bromide
C8H4BrCl2F3
MFCD11519349
69° to 72°C
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit