Alfa Aesar™ 2,6-Dichloro-4-fluoroaniline, 98+%

Manufacturer: Alfa Aesar™ B2544306

Quantity 5g

 View more versions of this product

Catalog No. AAB2544306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 344-19-4
    Molecular Formula C6H4Cl2FN
    Formula Weight 180.01
    MDL Number MFCD00142845
    UN Number UN2811
    Beilstein 2718192
    Chemical Name or Material 2,6-Dichloro-4-fluoroaniline

    Specifications

    Formula Weight 180.01
    Odor Recognizable
    MDL Number MFCD00142845
    Molecular Formula C6H4Cl2FN
    CAS 344-19-4
    UN Number UN2811
    Beilstein 2718192
    Melting Point 54° to 58°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H373-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Quantity 5g
    Chemical Name or Material 2,6-Dichloro-4-fluoroaniline
    Assay ≥98%

    Description

    Applications
    2,6-Dichloro-4-fluoroaniline is used as a pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

    GHS H Statement
    H301-H311-H373-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P301+P310-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit