Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dichloro-4-fluoroaniline, 98+%

Click to view available options
Quantity:
1g
5g
344-19-4
C6H4Cl2FN
180.003
MFCD00142845
YAUYKCFMKMZTEX-UHFFFAOYSA-N
2,6-dichloro-4-fluorobenzenamine,2,6-dichloro-4-fluoro-aniline,2',6'-dichloro-4'-fluoroaniline,benzenamine, 2,6-dichloro-4-fluoro,2,6-dichloro-4-fluorophenylamine,pubchem3451,acmc-1ad5c,ksc572g5t,2,6,-dichloro-4-fluoroaniline,2,6 dichloro-4-fluorophenylamine
2774008
2,6-dichloro-4-fluoroaniline
C1=C(C=C(C(=C1Cl)N)Cl)F
344-19-4
C6H4Cl2FN
180.003
MFCD00142845
YAUYKCFMKMZTEX-UHFFFAOYSA-N
2,6-dichloro-4-fluorobenzenamine,2,6-dichloro-4-fluoro-aniline,2',6'-dichloro-4'-fluoroaniline,benzenamine, 2,6-dichloro-4-fluoro,2,6-dichloro-4-fluorophenylamine,pubchem3451,acmc-1ad5c,ksc572g5t,2,6,-dichloro-4-fluoroaniline,2,6 dichloro-4-fluorophenylamine
2774008
2,6-dichloro-4-fluoroaniline
C1=C(C=C(C(=C1Cl)N)Cl)F
Applications
2,6-Dichloro-4-fluoroaniline is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Melting Point 54°C to 58°C
Odor Recognizable
Quantity 1g
UN Number UN2811
Beilstein 2718192
Formula Weight 180.01
Percent Purity ≥98%
Chemical Name or Material 2,6-Dichloro-4-fluoroaniline

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.