Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-4-isocyanatopyridine, 90%, Thermo Scientific™

Click to view available options
Quantity:
1g
10g
159178-03-7
C6H2Cl2N2O
189
OCDFIQXUCLQQHY-UHFFFAOYSA-N
2,6-dichloro-4-pyridyl isocyanate, 2,6-dichloropyridin-4-isocyanate, 2,6-dichloropyridin-4-yl isocyanate, pyridine,2,6-dichloro-4-isocyanato, pyridine, 2,6-dichloro-4-isocyanato, 2,6-dichloropyridin-4-ylisocyanate, pyridine, 2,6-dichloro-4-isocyanato-9ci
2781658
2,6-dichloro-4-isocyanatopyridine
C1=C(C=C(N=C1Cl)Cl)N=C=O
159178-03-7
C6H2Cl2N2O
189
OCDFIQXUCLQQHY-UHFFFAOYSA-N
2,6-dichloro-4-pyridyl isocyanate, 2,6-dichloropyridin-4-isocyanate, 2,6-dichloropyridin-4-yl isocyanate, pyridine,2,6-dichloro-4-isocyanato, pyridine, 2,6-dichloro-4-isocyanato, 2,6-dichloropyridin-4-ylisocyanate, pyridine, 2,6-dichloro-4-isocyanato-9ci
2781658
2,6-dichloro-4-isocyanatopyridine
C1=C(C=C(N=C1Cl)Cl)N=C=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 38.0°C to 51.0°C
Assay Percent Range 89% min. (GC)
Infrared Spectrum Authentic
Quantity 10g
Formula Weight 189
Percent Purity 90%
Chemical Name or Material 2,6-Dichloro-4-isocyanatopyridine

GHS H Statement
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause allergy or asthma sympto

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

Danger

RUO – Research Use Only