Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dichloro-4-nitroaniline, 95%

Click to view available options
Quantity:
25g
100g
500g
99-30-9
C6H4Cl2N2O2
207.01
MFCD00007677
BIXZHMJUSMUDOQ-UHFFFAOYSA-N
dichloran,dicloran,allisan,botran,dcna,ditranil,batran,bortran,resisan,dcna fungicide
7430
CHEBI:27864
2,6-dichloro-4-nitroaniline
C1=C(C=C(C(=C1Cl)N)Cl)[N+](=O)[O-]
99-30-9
C6H4Cl2N2O2
207.01
MFCD00007677
BIXZHMJUSMUDOQ-UHFFFAOYSA-N
dichloran,dicloran,allisan,botran,dcna,ditranil,batran,bortran,resisan,dcna fungicide
7430
CHEBI:27864
2,6-dichloro-4-nitroaniline
C1=C(C=C(C(=C1Cl)N)Cl)[N+](=O)[O-]
Applications
It is used as dyestuff, agricultural and pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Quantity 100g
Melting Point 185°C to 192°C
Boiling Point 130°C (2mmHg)
Odor Recognizable
Beilstein 1459581
Formula Weight 207.02
Percent Purity 95%
Chemical Name or Material 2,6-Dichloro-4-nitroaniline

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P262-P305+P351+P338
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : BX2975000

TSCA : TSCA