Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass Bottle
82671-02-1
C6HCl2FN2
190.99
DEDKKOOGYIMMBC-UHFFFAOYSA-N
2,6-dichloro-5-fluoro-3-pyridinecarbonitrile,2,6-dichloro-5-fluoronicotinonitrile,2,6-dichloro-3-cyano-5-fluoropyridine,3-cyano-2,6-dichloro-5-fluoropyridine,2,6-dichloro-5-fluoro-nicotinonitrile,2,6-dichloro-5-fluoro-3-cyanopyridine,2,6-dichloro-5-fluoro-pyridine-3-carbonitrile,3-pyridinecarbonitrile, 2,6-dichloro-5-fluoro,pubchem10945,acmc-209ppr
2733849
2,6-dichloro-5-fluoropyridine-3-carbonitrile
C1=C(C(=NC(=C1F)Cl)Cl)C#N
82671-02-1
C6HCl2FN2
190.99
DEDKKOOGYIMMBC-UHFFFAOYSA-N
2,6-dichloro-5-fluoro-3-pyridinecarbonitrile,2,6-dichloro-5-fluoronicotinonitrile,2,6-dichloro-3-cyano-5-fluoropyridine,3-cyano-2,6-dichloro-5-fluoropyridine,2,6-dichloro-5-fluoro-nicotinonitrile,2,6-dichloro-5-fluoro-3-cyanopyridine,2,6-dichloro-5-fluoro-pyridine-3-carbonitrile,3-pyridinecarbonitrile, 2,6-dichloro-5-fluoro,pubchem10945,acmc-209ppr
2733849
2,6-dichloro-5-fluoropyridine-3-carbonitrile
C1=C(C(=NC(=C1F)Cl)Cl)C#N
Packaging Glass Bottle
Quantity 100g
Color Yellow
Melting Point 85.0°C to 92.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Formula Weight 190.99
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 2, 6-Dichloro-5-fluoro-3-pyridinecarbonitrile

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning