Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-5-fluoronicotinoyl chloride, 90%, Acros Organics

Click to view available options
Quantity:
5g
96568-02-4
C6HCl3FNO
228.428
AATVXELAYCLVTJ-UHFFFAOYSA-N
2,6-dichloro-5-fluoronicotinoyl chloride,2,6-dichloro-5-fluoro-3-pyridinecarbonyl chloride,2,6-dichloro-5-fluoro-pyridine-3-carbonyl chloride,3-pyridinecarbonylchloride, 2,6-dichloro-5-fluoro,2,6-dichloro-5-fluoro-nicotinoylchloride,2,6-dichloro-5-fluoronicotinic acid chloride,3-pyridinecarbonyl chloride, 2,6-dichloro-5-fluoro,2,6-dichloro-5-fluoropyridine-3-carboxylic acid chloride,2,6-bis chloranyl-5-fluoranyl-pyridine-3-carbonyl chloride
2733666
2,6-dichloro-5-fluoropyridine-3-carbonyl chloride
C1=C(C(=NC(=C1F)Cl)Cl)C(=O)Cl
96568-02-4
C6HCl3FNO
228.428
AATVXELAYCLVTJ-UHFFFAOYSA-N
2,6-dichloro-5-fluoronicotinoyl chloride,2,6-dichloro-5-fluoro-3-pyridinecarbonyl chloride,2,6-dichloro-5-fluoro-pyridine-3-carbonyl chloride,3-pyridinecarbonylchloride, 2,6-dichloro-5-fluoro,2,6-dichloro-5-fluoro-nicotinoylchloride,2,6-dichloro-5-fluoronicotinic acid chloride,3-pyridinecarbonyl chloride, 2,6-dichloro-5-fluoro,2,6-dichloro-5-fluoropyridine-3-carboxylic acid chloride,2,6-bis chloranyl-5-fluoranyl-pyridine-3-carbonyl chloride
2733666
2,6-dichloro-5-fluoropyridine-3-carbonyl chloride
C1=C(C(=NC(=C1F)Cl)Cl)C(=O)Cl
Infrared Spectrum Authentic
Quantity 5g
Refractive Index 1.5680 to 1.5730 (20°C, 589nm)
Color Yellow
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Formula Weight 228.44
Physical Form Liquid
Percent Purity ≥88% (acid halide)
Chemical Name or Material 2, 6-Dichloro-5-fluoronicotinoyl chloride, 90%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE