Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-5-fluoropyridine-3-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
82671-06-5
C6H2Cl2FNO2
209.985
MFCD00799517
LTDGKGCHRNNCAC-UHFFFAOYSA-N
2,6-dichloro-5-fluoronicotinic acid,2,6-dichloro-5-fluoro-nicotinic acid,2,6-dichloro-5-fluoro-3-pyridinecarboxylic acid,2,6-dichloro-5-fluoro-pyridine-3-carboxylic acid,2,6-dichloro-5-fluoro nicotinic acid,3-pyridinecarboxylic acid, 2,6-dichloro-5-fluoro,pubchem10943,acmc-1bluq,ksc448a5j,2,6-dichloro-5-fluoro-nicotinicacid
2733659
2,6-dichloro-5-fluoropyridine-3-carboxylic acid
C1=C(C(=NC(=C1F)Cl)Cl)C(=O)O
82671-06-5
C6H2Cl2FNO2
209.985
MFCD00799517
LTDGKGCHRNNCAC-UHFFFAOYSA-N
2,6-dichloro-5-fluoronicotinic acid,2,6-dichloro-5-fluoro-nicotinic acid,2,6-dichloro-5-fluoro-3-pyridinecarboxylic acid,2,6-dichloro-5-fluoro-pyridine-3-carboxylic acid,2,6-dichloro-5-fluoro nicotinic acid,3-pyridinecarboxylic acid, 2,6-dichloro-5-fluoro,pubchem10943,acmc-1bluq,ksc448a5j,2,6-dichloro-5-fluoro-nicotinicacid
2733659
2,6-dichloro-5-fluoropyridine-3-carboxylic acid
C1=C(C(=NC(=C1F)Cl)Cl)C(=O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Melting Point 153°C to 158°C
Color Yellow to Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 209.99
Physical Form Fine Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2, 6-Dichloro-5-fluoropyridine-3-carboxylic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning