Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',6'-Dichloroacetophenone, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2040-05-3
C8H6Cl2O
189.04
MFCD00016339
HYBDSXBLGCQKRE-UHFFFAOYSA-N
2',6'-dichloroacetophenone,1-2,6-dichlorophenyl ethanone,2,6-dichloroacetophenone,1-2,6-dichlorophenyl ethan-1-one,ethanone, 1-2,6-dichlorophenyl,2,6-dichloro acetophenone,1-acetyl-2,6-dichlorobenzene,zlchem 1084,pubchem12362,acmc-1cobf
74877
1-(2,6-dichlorophenyl)ethanone
CC(=O)C1=C(C=CC=C1Cl)Cl
2040-05-3
C8H6Cl2O
189.04
MFCD00016339
HYBDSXBLGCQKRE-UHFFFAOYSA-N
2',6'-dichloroacetophenone,1-2,6-dichlorophenyl ethanone,2,6-dichloroacetophenone,1-2,6-dichlorophenyl ethan-1-one,ethanone, 1-2,6-dichlorophenyl,2,6-dichloro acetophenone,1-acetyl-2,6-dichlorobenzene,zlchem 1084,pubchem12362,acmc-1cobf
74877
1-(2,6-dichlorophenyl)ethanone
CC(=O)C1=C(C=CC=C1Cl)Cl
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color Colorless
Melting Point 41.0°C to 42.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 189.04
Physical Form Low Melting Solid
Percent Purity 97%
Chemical Name or Material 2', 6'-Dichloroacetophenone
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-035-7