Alfa Aesar™ 2,6-Dichloroaniline, 98%

Manufacturer: Alfa Aesar™ A1232014

Quantity 25g

 View more versions of this product

Catalog No. AAA1232014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 608-31-1
    Molecular Formula C6H5Cl2N
    Formula Weight 162.02
    MDL Number MFCD00007675
    UN Number UN3442
    Beilstein 775084
    Chemical Name or Material 2,6-Dichloroaniline

    Specifications

    Formula Weight 162.02
    Health Hazard 3 GHS P Statement
    P260-P261-P301+P310-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 775084
    Health Hazard 1 Danger
    Boiling Point 97°C (5mmHg)
    Melting Point 37° to 40°C
    Quantity 25g
    TSCA TSCA
    Molecular Formula C6H5Cl2N
    MDL Number MFCD00007675
    CAS 608-31-1
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H373
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
    Flash Point 118°C (244°F)
    UN Number UN3442
    RTECS Number CX9862655
    Chemical Name or Material 2,6-Dichloroaniline
    Assay 98%

    Description

    Applications
    2,6-Dichloroaniline is used in the pharmaceutical industry for the synthesis of antibacterial agents of lomefloxacin, Lee uric acid and clonidine.

    Solubility
    Soluble in water (1.6 g/L at 20°C), dimethyl sulfoxide and methanol.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H311-H331-H373
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P261-P301+P310-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit