Alfa Aesar™ 2,6-Dichloroanisole, 99%

Manufacturer: Alfa Aesar™ B2530209

Quantity 10g

 View more versions of this product

Catalog No. AAB2530209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1984-65-2
    Molecular Formula C7H6Cl2O
    Formula Weight 177.03
    MDL Number MFCD00000575
    Synonym 1,3-Dichloro-2-methoxybenzene
    Chemical Name or Material 2,6-Dichloroanisole

    Specifications

    Formula Weight 177.03
    Melting Point 10° to 11°C
    Quantity 10g
    TSCA TSCA
    Molecular Formula C7H6Cl2O
    MDL Number MFCD00000575
    CAS 1984-65-2
    Synonym 1,3-Dichloro-2-methoxybenzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 91°C (196°F)
    Boiling Point 117°C (20mmHg)
    Chemical Name or Material 2,6-Dichloroanisole
    Assay 99%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit