Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzoxazole 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
3621-82-7
C7H3Cl2NO
188.007
MFCD07368635
LVVQTPZQNHQLOM-UHFFFAOYSA-N
77175
2,6-dichloro-1,3-benzoxazole
C1=CC2=C(C=C1Cl)OC(=N2)Cl
3621-82-7
C7H3Cl2NO
188.007
MFCD07368635
LVVQTPZQNHQLOM-UHFFFAOYSA-N
77175
2,6-dichloro-1,3-benzoxazole
C1=CC2=C(C=C1Cl)OC(=N2)Cl
Color White-Yellow
Melting Point 50°C
Quantity 5g
Boiling Point 110°C
Formula Weight 188.01
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,6-Dichlorobenzoxazole