Alfa Aesar™ 2,6-Dichlorobenzyl alcohol, 99%

Manufacturer: Alfa Aesar™ A1219618

Quantity 50g

 View more versions of this product

Catalog No. AAA1219618

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 15258-73-8
    Molecular Formula C7H6Cl2O
    Formula Weight 177.03
    MDL Number MFCD00004609
    Beilstein 1938356
    Chemical Name or Material 2,6-Dichlorobenzyl alcohol

    Specifications

    Formula Weight 177.03
    CAS 15258-73-8
    Boiling Point 150°C (25mmHg)
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 50g
    Molecular Formula C7H6Cl2O
    MDL Number MFCD00004609
    Beilstein 1938356
    Melting Point 95° to 99°C
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    RTECS Number DO0900000
    Chemical Name or Material 2,6-Dichlorobenzyl alcohol
    Assay 99%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit