Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzyl Alcohol 99%, Acros Organics

Click to view available options
Quantity:
25g
15258-73-8
C7H6Cl2O
177.024
MFCD00004609
WKKHCCZLKYKUDN-UHFFFAOYSA-N
2,6-dichlorobenzyl alcohol,2,6-dichlorophenyl methanol,benzenemethanol, 2,6-dichloro,2,6-dichlorobenzenemethanol,benzyl alcohol, 2,6-dichloro,2,6-dcal,2,6-dichlorobenzylalcohol,benzenemethanol, 2,6-dichloro-9ci,2,6-dichloro-benzenemethanol,2,6-dichloro-phenyl-methanol
27156
(2,6-dichlorophenyl)methanol
C1=CC(=C(C(=C1)Cl)CO)Cl
15258-73-8
C7H6Cl2O
177.024
MFCD00004609
WKKHCCZLKYKUDN-UHFFFAOYSA-N
2,6-dichlorobenzyl alcohol,2,6-dichlorophenyl methanol,benzenemethanol, 2,6-dichloro,2,6-dichlorobenzenemethanol,benzyl alcohol, 2,6-dichloro,2,6-dcal,2,6-dichlorobenzylalcohol,benzenemethanol, 2,6-dichloro-9ci,2,6-dichloro-benzenemethanol,2,6-dichloro-phenyl-methanol
27156
(2,6-dichlorophenyl)methanol
C1=CC(=C(C(=C1)Cl)CO)Cl
Quantity 25g
Color White
Infrared Spectrum Authentic
Melting Point 95°C to 98°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula Cl2C6H3CH2OH
Formula Weight 177.03
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2, 6-Dichlorobenzyl alcohol, 99%
Show More Show Less