Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (2,6-Dichlorobenzyl)triphenylphosphonium chloride, 98%

Click to view available options
Quantity:
5g
25g
100g
18583-54-5
C25H20Cl3P
457.759
MFCD00061392
NBWPWZJEEQWREI-UHFFFAOYSA-M
2,6-dichlorobenzyl triphenylphosphonium chloride,2,6-dichlorophenyl methyl triphenylphosphanium chloride,acmc-20ao99,2,6-dichlorobenzyltriphenylphosphoniumchloride,2,6-dichlorophenyl methyl-triphenyl-phosphonium chloride,2,6-dichlorophenyl methyl triphenyl phosphanium chloride,phosphonium, 2,6-dichlorophenyl methyl triphenyl-, chloride 1:1
57370833
(2,6-dichlorophenyl)methyl-triphenylphosphanium;chloride
C1=CC=C(C=C1)[P+](CC2=C(C=CC=C2Cl)Cl)(C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]
18583-54-5
C25H20Cl3P
457.759
MFCD00061392
NBWPWZJEEQWREI-UHFFFAOYSA-M
2,6-dichlorobenzyl triphenylphosphonium chloride,2,6-dichlorophenyl methyl triphenylphosphanium chloride,acmc-20ao99,2,6-dichlorobenzyltriphenylphosphoniumchloride,2,6-dichlorophenyl methyl-triphenyl-phosphonium chloride,2,6-dichlorophenyl methyl triphenyl phosphanium chloride,phosphonium, 2,6-dichlorophenyl methyl triphenyl-, chloride 1:1
57370833
(2,6-dichlorophenyl)methyl-triphenylphosphanium;chloride
C1=CC=C(C=C1)[P+](CC2=C(C=CC=C2Cl)Cl)(C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]
Quantity 5g
Formula Weight 457.78
Percent Purity 98%
Chemical Name or Material (2,6-Dichlorobenzyl)triphenylphosphonium chloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures