Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloroindophenol, Sodium Derivative, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
5g
10g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

620-45-1
C12H6Cl2NNaO2
290.075
CVSUAFOWIXUYQA-UHFFFAOYSA-M
2,6-Dichlorophenalindephenol Sodium Salt
23697355
CHEBI:948

High quality chemicals for laboratory and specialized industrial use

Quantity 5g
Packaging Wide Mouth Amber Glass Bottle
SMILES C1=CC(=CC=C1N=C2C=C(C(=O)C(=C2)Cl)Cl)[O-].[Na+]
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
Formula Weight 290.08
Chemical Name or Material 2,6-Dichloroindophenol, Sodium Derivative
Grade Baker Analyzed™ ACS Reagent