Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloroindophenol, sodium salt hydrate, 98+%, Acros Organics™

Click to view available options
Quantity:
1g
10g
25g
100g
Packaging:
Glass bottle
1266615-56-8
C12H10Cl2NNaO4
326.105
WZHXLAHEFATELD-UHFFFAOYSA-M
Tillman's reagent hydrate
23696612
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate;dihydrate
C1=CC(=CC=C1N=C2C=C(C(=O)C(=C2)Cl)Cl)[O-].O.O.[Na+]
1266615-56-8
C12H10Cl2NNaO4
326.105
WZHXLAHEFATELD-UHFFFAOYSA-M
Tillman's reagent hydrate
23696612
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate;dihydrate
C1=CC(=CC=C1N=C2C=C(C(=O)C(=C2)Cl)Cl)[O-].O.O.[Na+]
Assay Percent Range 98+%
Infrared Spectrum Authentic
Loss on Drying 14% max. (1 g, 120°C)
Formula Weight 290.07
Physical Form Powder
Percent Purity ≥98%
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: freely soluble in alcohol
Suitability For determination of ascorbic acid: Passes Test
Color Brown to Red
Quantity 100g
Chemical Name or Material 2, 6-Dichloroindophenol, sodium salt hydrate
Show More Show Less