Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ 2,6-Dichloroindophenol sodium salt hydrate, Honeywell Fluka™

Click to view available options
Quantity:
1g
5g
25g
620-45-1
C12H6Cl2NNaO2
290.075
MFCD00150014
CVSUAFOWIXUYQA-UHFFFAOYSA-M
2,6-dichloroindophenol sodium salt,2,6-dichlorophenolindophenol sodium salt,tillman's reagent,tillman's reagenz,dichlorphenol-indophenolnatrium,2,6-dichloroindophenol sodium,unii-kad7q8xo1y,sodium 2,6-dichloroindophenol,sodium 2,6-dichloroindophenolate,2,6-dichloroindophenol, sodium salt
23697355
CHEBI:948
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
C1=CC(=CC=C1N=C2C=C(C(=O)C(=C2)Cl)Cl)[O-].[Na+]
620-45-1
C12H6Cl2NNaO2
290.075
MFCD00150014
CVSUAFOWIXUYQA-UHFFFAOYSA-M
2,6-dichloroindophenol sodium salt,2,6-dichlorophenolindophenol sodium salt,tillman's reagent,tillman's reagenz,dichlorphenol-indophenolnatrium,2,6-dichloroindophenol sodium,unii-kad7q8xo1y,sodium 2,6-dichloroindophenol,sodium 2,6-dichloroindophenolate,2,6-dichloroindophenol, sodium salt
23697355
CHEBI:948
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
C1=CC(=CC=C1N=C2C=C(C(=O)C(=C2)Cl)Cl)[O-].[Na+]

≥97.0% (calc. based on dry substance, AT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Quantity 1g
UN Number NONH for all modes of transport
Beilstein 3641229
Formula Weight 290.08g/mol (anhydrous basis)
Percent Purity ≥97% (calculated based on dry substance, AT)
Chemical Name or Material 2,6-Dichloroindophenol sodium salt hydrate

RTECSNumber : GU5495000