Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloroisonicotinic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
5398-44-7
C6H3Cl2NO2
191.995
MFCD00234147
SQSYNRCXIZHKAI-UHFFFAOYSA-N
2,6-dichloroisonicotinic acid,2,6-dichloro-isonicotinic acid,2,6-dichloro-4-pyridinecarboxylic acid,4-pyridinecarboxylic acid, 2,6-dichloro,2,6-dichloroisonicotinicacid,2,6-dichloro-4-pyridinyl carboxylic acid,pyridine-4-carboxylic acid, 2,6-dichloro,2,6-dichlorisonicotins,enamine_005327,zlchem 403
94830
CHEBI:73179
2,6-dichloropyridine-4-carboxylic acid
C1=C(C=C(N=C1Cl)Cl)C(=O)O
5398-44-7
C6H3Cl2NO2
191.995
MFCD00234147
SQSYNRCXIZHKAI-UHFFFAOYSA-N
2,6-dichloroisonicotinic acid,2,6-dichloro-isonicotinic acid,2,6-dichloro-4-pyridinecarboxylic acid,4-pyridinecarboxylic acid, 2,6-dichloro,2,6-dichloroisonicotinicacid,2,6-dichloro-4-pyridinyl carboxylic acid,pyridine-4-carboxylic acid, 2,6-dichloro,2,6-dichlorisonicotins,enamine_005327,zlchem 403
94830
CHEBI:73179
2,6-dichloropyridine-4-carboxylic acid
C1=C(C=C(N=C1Cl)Cl)C(=O)O
Infrared Spectrum Authentic
Quantity 1g
Color Beige to White or Brown
Packaging Glass bottle
Melting Point 206°C to 210°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 192
Physical Form Powder and/or Chunks
Percent Purity ≥97.5%
Chemical Name or Material 2, 6-Dichloroisonicotinic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning