Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorophenylboronic acid, 97%

Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
73852-17-2
C6H5BCl2O2
190.814
MFCD00064869
CXDPUSMFYPQXCV-UHFFFAOYSA-N
2,6-dichlorobenzeneboronic acid,2,6-dichlorophenyl boronic acid,2,6-dichlorophenylboronicacid,2,6-dichlorophenyl boranediol,boronic acid, 2,6-dichlorophenyl,pubchem7938,acmc-1bc3m,ksc489k8l,2,6-dichloro-phenylboronic acid
2734332
(2,6-dichlorophenyl)boronic acid
B(C1=C(C=CC=C1Cl)Cl)(O)O
73852-17-2
C6H5BCl2O2
190.814
MFCD00064869
CXDPUSMFYPQXCV-UHFFFAOYSA-N
2,6-dichlorobenzeneboronic acid,2,6-dichlorophenyl boronic acid,2,6-dichlorophenylboronicacid,2,6-dichlorophenyl boranediol,boronic acid, 2,6-dichlorophenyl,pubchem7938,acmc-1bc3m,ksc489k8l,2,6-dichloro-phenylboronic acid
2734332
(2,6-dichlorophenyl)boronic acid
B(C1=C(C=CC=C1Cl)Cl)(O)O
Infrared Spectrum Authentic
Color White
Quantity 250mg
Packaging Amber Glass Bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 190.82
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 6-Dichlorophenylboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.