Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorophenylboronic acid, 98%, Thermo Scientific™

Catalog Number AC359115000
Click to view available options
Quantity:
500 mg
Packaging:
Glass Bottle
73852-17-2
C6H5BCl2O2
190.81
MFCD00064869
CXDPUSMFYPQXCV-UHFFFAOYSA-N
2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, 2,6-dichlorophenyl boranediol, boronic acid, 2,6-dichlorophenyl, pubchem7938, acmc-1bc3m, ksc489k8l, 2,6-dichloro-phenylboronic acid
2734332
(2,6-dichlorophenyl)boronic acid
OB(O)C1=C(Cl)C=CC=C1Cl
73852-17-2
C6H5BCl2O2
190.81
MFCD00064869
CXDPUSMFYPQXCV-UHFFFAOYSA-N
2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, 2,6-dichlorophenyl boranediol, boronic acid, 2,6-dichlorophenyl, pubchem7938, acmc-1bc3m, ksc489k8l, 2,6-dichloro-phenylboronic acid
2734332
(2,6-dichlorophenyl)boronic acid
OB(O)C1=C(Cl)C=CC=C1Cl
CAS Min % 97.5
CAS Max % 100.0
Melting Point 153°C to 154°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 500 mg
Formula Weight 190.82
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 6-Dichlorophenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit