Alfa Aesar™ 2,6-Dichlorophenylhydrazine hydrochloride, 98+%

Manufacturer: Alfa Aesar™ A1196814

Quantity 25g

 View more versions of this product

Catalog No. AAA1196814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 50709-36-9
    Molecular Formula C6H6Cl2N2·HCl
    Formula Weight 213.5
    MDL Number MFCD00012930
    Beilstein 4569738
    Chemical Name or Material 2,6-Dichlorophenylhydrazine hydrochloride

    Specifications

    Formula Weight 213.5
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H6Cl2N2·HCl
    CAS 50709-36-9
    MDL Number MFCD00012930
    Beilstein 4569738
    Health Hazard 1 Danger
    Melting Point ≈225°C (dec.)
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H301-H311-H332-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.
    Chemical Name or Material 2,6-Dichlorophenylhydrazine hydrochloride
    Assay ≥98%

    GHS H Statement
    H301-H311-H332-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit