Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloropurine, 97%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
5451-40-1
C5H2Cl2N4
189.00
MFCD00077725
RMFWVOLULURGJI-UHFFFAOYSA-N
2,6-dichloropurine, 2,6-dichloro-9h-purine, 2,6-dichloro-1h-purine, 9h-purine, 2,6-dichloro, 2,6-dichloro purine, 2,6 dichloropurine, 2,6-dichioropurine, purine,6-dichloro, zlchem 226, 2,6-dichloro-purine
5324412
2,6-dichloro-7H-purine
ClC1=NC(Cl)=C2NC=NC2=N1
5451-40-1
C5H2Cl2N4
189.00
MFCD00077725
RMFWVOLULURGJI-UHFFFAOYSA-N
2,6-dichloropurine, 2,6-dichloro-9h-purine, 2,6-dichloro-1h-purine, 9h-purine, 2,6-dichloro, 2,6-dichloro purine, 2,6 dichloropurine, 2,6-dichioropurine, purine,6-dichloro, zlchem 226, 2,6-dichloro-purine
5324412
2,6-dichloro-7H-purine
ClC1=NC(Cl)=C2NC=NC2=N1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Melting Point 184.0°C to 186.0°C
Assay Percent Range 96% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250mg
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone and ethyl acetate
Formula Weight 189
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloropurine, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-681-6

TSCA : TSCA

RUO – Research Use Only