Alfa Aesar™ 2,6-Dichloropyridine, 98%

Manufacturer: Alfa Aesar™ A113640E

Quantity 2500g

 View more versions of this product

Catalog No. AAA113640E

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2402-78-0
    Molecular Formula C5H3Cl2N
    Formula Weight 147.99
    MDL Number MFCD00006244
    UN Number UN2811
    Beilstein 108664
    Chemical Name or Material 2,6-Dichloropyridine

    Specifications

    Formula Weight 147.99
    Health Hazard 3 GHS P Statement
    P261-P280h-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    MDL Number MFCD00006244
    CAS 2402-78-0
    UN Number UN2811
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Melting Point 84° to 88°C
    Quantity 2500g
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number US8400000
    Odor Unpleasant
    TSCA TSCA
    Molecular Formula C5H3Cl2N
    Beilstein 108664
    Boiling Point 211°C
    Chemical Name or Material 2,6-Dichloropyridine
    Assay 98%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280h-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit