Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloropyrimidine-4-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
16492-28-7
C5H2Cl2N2O2
192.99
MOXUTXQEKYPKKS-UHFFFAOYSA-N
2,4-dichloropyrimidine-6-carboxylic acid,4-pyrimidinecarboxylic acid, 2,6-dichloro,2,6-dichloro-pyrimidine-4-carboxylic acid,4-pyrimidinecarboxylicacid, 2,6-dichloro,2,4-dichloropyrimidin-6-yl carboxylic acid,2,6-dichloro-4-pyrimidine carboxylic acid,2 pound not6-dichloropyrimidine-4-carboxylicacid,2,6-dichloropyrimidine-4-carboxylic acid
240530
2,6-dichloropyrimidine-4-carboxylic acid
C1=C(N=C(N=C1Cl)Cl)C(=O)O
16492-28-7
C5H2Cl2N2O2
192.99
MOXUTXQEKYPKKS-UHFFFAOYSA-N
2,4-dichloropyrimidine-6-carboxylic acid,4-pyrimidinecarboxylic acid, 2,6-dichloro,2,6-dichloro-pyrimidine-4-carboxylic acid,4-pyrimidinecarboxylicacid, 2,6-dichloro,2,4-dichloropyrimidin-6-yl carboxylic acid,2,6-dichloro-4-pyrimidine carboxylic acid,2 pound not6-dichloropyrimidine-4-carboxylicacid,2,6-dichloropyrimidine-4-carboxylic acid
240530
2,6-dichloropyrimidine-4-carboxylic acid
C1=C(N=C(N=C1Cl)Cl)C(=O)O
Melting Point 85°C to 95°C
Quantity 5g
Assay Percent Range 97%
Formula Weight 192.99
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloropyrimidine-4-carboxylic acid

GHS Signal Word: Warning