Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloroquinone-4-chloroimide, 95%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
1 kg
Packaging:
Amber Glass Bottle
101-38-2
C6H2Cl3NO
210.44
YHUMTHWQGWPJOQ-UHFFFAOYSA-N
2,6-dichloro-4-(chloroimino)cyclohexa-2,5-dien-1-one
ClN=C1C=C(Cl)C(=O)C(Cl)=C1
101-38-2
C6H2Cl3NO
210.44
YHUMTHWQGWPJOQ-UHFFFAOYSA-N
2,6-dichloro-4-(chloroimino)cyclohexa-2,5-dien-1-one
ClN=C1C=C(Cl)C(=O)C(Cl)=C1

D2746, 101-38-2, C6H2Cl3NO

Spectrum™ 2,6-Dichloroquinone-4-chloroimide
Packaging Amber Glass Bottle
Quantity 1 g
CAS Max % 1
UN Number UN3224
Percent Purity 95%