Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloroquinone-4-chloroimide 97.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
25g
101-38-2
C6H2Cl3NO
210.438
MFCD00001611
YHUMTHWQGWPJOQ-UHFFFAOYSA-N
2,6-dichloroquinone-4-chloroimide,gibbs reagent,2,6-dichloro-4-chloroimino cyclohexa-2,5-dienone,2,5-cyclohexadien-1-one, 2,6-dichloro-4-chloroimino,2,6-dichloroquinonechloroimine,2,6-dichloroquinone chloroimide,2,6-dichloroquinone-4-chloroimine,2,6-dichloro-4-n-chloroquinonimine,2,6-dichlorobenzoquinone chloroimide,p-benzoquinone imine, n,2,6-trichloro
7556
2,6-dichloro-4-chloroiminocyclohexa-2,5-dien-1-one
C1=C(C(=O)C(=CC1=NCl)Cl)Cl
101-38-2
C6H2Cl3NO
210.438
MFCD00001611
YHUMTHWQGWPJOQ-UHFFFAOYSA-N
2,6-dichloroquinone-4-chloroimide,gibbs reagent,2,6-dichloro-4-chloroimino cyclohexa-2,5-dienone,2,5-cyclohexadien-1-one, 2,6-dichloro-4-chloroimino,2,6-dichloroquinonechloroimine,2,6-dichloroquinone chloroimide,2,6-dichloroquinone-4-chloroimine,2,6-dichloro-4-n-chloroquinonimine,2,6-dichlorobenzoquinone chloroimide,p-benzoquinone imine, n,2,6-trichloro
7556
2,6-dichloro-4-chloroiminocyclohexa-2,5-dien-1-one
C1=C(C(=O)C(=CC1=NCl)Cl)Cl
Color Undesignated
Melting Point 65°C
Quantity 25g
UN Number 1479
Formula Weight 210.44
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material 2,6-Dichloroquinone-4-chloroimide

RTECSNumber : GU5470000

TSCA : Yes