Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dichlorothiobenzamide, 97%

Manufacturer:  Alfa Aesar™ H2663603

Quantity 1g

 View more versions of this product

Catalog No. AAH2663603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1918-13-4
206.09
1910353
2,6-Dichlorothiobenzamide
C7H5Cl2NS
MFCD00053523
Chlorothiamide

Specifications

1g
C7H5Cl2NS
MFCD00053523
149° to 153°C
Chlorothiamide
97%
1918-13-4
206.09
1910353
>100°C (212°F)
2,6-Dichlorothiobenzamide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CV3850000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit