Alfa Aesar™ 2,6-Dichlorothiobenzamide, 97%

Manufacturer: Alfa Aesar™ H2663606

Quantity 5g

 View more versions of this product

Catalog No. AAH2663606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1918-13-4
    Molecular Formula C7H5Cl2NS
    Formula Weight 206.09
    MDL Number MFCD00053523
    Beilstein 1910353
    Synonym Chlorothiamide
    Chemical Name or Material 2,6-Dichlorothiobenzamide

    Specifications

    Formula Weight 206.09
    Quantity 5g
    Molecular Formula C7H5Cl2NS
    Beilstein 1910353
    Melting Point 149° to 153°C
    Flash Point >100°C (212°F)
    MDL Number MFCD00053523
    CAS 1918-13-4
    Synonym Chlorothiamide
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number CV3850000
    Chemical Name or Material 2,6-Dichlorothiobenzamide
    Assay 97%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit