Alfa Aesar™ 2,6-Dichlorotoluene, 99%

Manufacturer: Alfa Aesar™ B2446336

Quantity 500g

 View more versions of this product

Catalog No. AAB2446336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 118-69-4
    Molecular Formula C7H6Cl2
    Formula Weight 161.03
    MDL Number MFCD00000576
    UN Number UN3082
    Beilstein 1907194
    Chemical Name or Material 2,6-Dichlorotoluene

    Specifications

    Formula Weight 161.03
    Molecular Formula C7H6Cl2
    MDL Number MFCD00000576
    Flash Point 82°C (179°F)
    Quantity 500g
    TSCA TSCA
    CAS 118-69-4
    UN Number UN3082
    Boiling Point 196° to 203°C
    Melting Point 2°C
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    Beilstein 1907194
    Chemical Name or Material 2,6-Dichlorotoluene
    Assay 99%

    Description

    Applications
    2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

    Solubility
    Soluble in water.

    Notes
    Store at room temperature. Incompatible with oxidizing agents.

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit