Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dichlorotoluene, 99%

Click to view available options
Quantity:
25g
100g
500g
118-69-4
C7H6Cl2
161.025
MFCD00000576
DMEDNTFWIHCBRK-UHFFFAOYSA-N
2,6-dichlorotoluene,benzene, 1,3-dichloro-2-methyl,toluene, 2,6-dichloro,unii-rvm09757ww,1,3-dichloro-2-methyl-benzene,toluene,6-dichloro,pubchem3701,2,6-dichloro-toluene,benzene,dichloromethyl,2,6-dichloro toluene
8368
1,3-dichloro-2-methylbenzene
CC1=C(C=CC=C1Cl)Cl
118-69-4
C7H6Cl2
161.025
MFCD00000576
DMEDNTFWIHCBRK-UHFFFAOYSA-N
2,6-dichlorotoluene,benzene, 1,3-dichloro-2-methyl,toluene, 2,6-dichloro,unii-rvm09757ww,1,3-dichloro-2-methyl-benzene,toluene,6-dichloro,pubchem3701,2,6-dichloro-toluene,benzene,dichloromethyl,2,6-dichloro toluene
8368
1,3-dichloro-2-methylbenzene
CC1=C(C=CC=C1Cl)Cl
Applications
2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Boiling Point 196°C to 203°C
Melting Point 2°C
Flash Point 82°C (179°F)
Quantity 25g
UN Number UN3082
Beilstein 1907194
Formula Weight 161.03
Percent Purity 99%
Chemical Name or Material 2,6-Dichlorotoluene

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : TSCA