Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Difluoro-3-methylaniline, 97%

Manufacturer:  Alfa Aesar™ H2665603

Quantity 1g

 View more versions of this product

Catalog No. AAH2665603


  Check Availability
Add to cart

Specifications

Description

Applications
2,6-Difluoro-3-methylaniline is used as a solvent is convenient as the pyridine can serve both as the solvent and the catalyst in the transformation.

Solubility
Soluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

144851-63-8
143.14
UN2810
C7H7F2N
MFCD06660182
2,6-Difluoro-3-methylaniline

Specifications

1g
C7H7F2N
MFCD06660182
2,6-Difluoro-3-methylaniline
144851-63-8
143.14
UN2810
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit