Alfa Aesar™ 2,6-Difluoro-3-methylanisole, 97%

Manufacturer: Alfa Aesar™ H2672206

Quantity 5g

 View more versions of this product

Catalog No. AAH2672206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 261763-33-1
    Molecular Formula C8H8F2O
    Formula Weight 158.15
    MDL Number MFCD01631341
    Chemical Name or Material 2,6-Difluoro-3-methylanisole

    Specifications

    Formula Weight 158.15
    Molecular Formula C8H8F2O
    MDL Number MFCD01631341
    CAS 261763-33-1
    Boiling Point 53°C (1mmHg)
    Quantity 5g
    Chemical Name or Material 2,6-Difluoro-3-methylanisole
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit