Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Difluoro-3-methylbenzaldehyde, 97+%

Click to view available options
Quantity:
1g
5g
261763-34-2
C8H6F2O
156.132
MFCD01631333
MXZXYEFIOADJMW-UHFFFAOYSA-N
2,6-difluoro-m-tolualdehyde,2,6-difluoro-3-methyl-benzaldehyde,benzaldehyde,2,6-difluoro-3-methyl,pubchem7147,acmc-1cmpd,benzaldehyde, 2,6-difluoro-3-methyl,2,6-bis fluoranyl-3-methyl-benzaldehyde,2,6-difluoro-3-methylbenzaldehyde,benzaldehyde, 2,6-difluoro-3-methyl-9ci
2774137
2,6-difluoro-3-methylbenzaldehyde
CC1=C(C(=C(C=C1)F)C=O)F
261763-34-2
C8H6F2O
156.132
MFCD01631333
MXZXYEFIOADJMW-UHFFFAOYSA-N
2,6-difluoro-m-tolualdehyde,2,6-difluoro-3-methyl-benzaldehyde,benzaldehyde,2,6-difluoro-3-methyl,pubchem7147,acmc-1cmpd,benzaldehyde, 2,6-difluoro-3-methyl,2,6-bis fluoranyl-3-methyl-benzaldehyde,2,6-difluoro-3-methylbenzaldehyde,benzaldehyde, 2,6-difluoro-3-methyl-9ci
2774137
2,6-difluoro-3-methylbenzaldehyde
CC1=C(C(=C(C=C1)F)C=O)F
Quantity 5g
Formula Weight 156.13
Percent Purity ≥97%
Chemical Name or Material 2,6-Difluoro-3-methylbenzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning