Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Difluoro-3-methylbenzoic acid, 97+%

Click to view available options
Quantity:
1g
5g
32890-88-3
C8H6F2O2
172.131
MFCD01631332
QZIVNVIPFGKDQE-UHFFFAOYSA-N
2,6-difluoro-3-methyl-benzoic acid,acmc-1af95,3-carboxy-2,4-difluorotoluene,benzoic acid,2,6-difluoro-3-methyl,2,6-difluoro-3-methylbenzoic acid 1g
2774142
2,6-difluoro-3-methylbenzoic acid
CC1=C(C(=C(C=C1)F)C(=O)O)F
32890-88-3
C8H6F2O2
172.131
MFCD01631332
QZIVNVIPFGKDQE-UHFFFAOYSA-N
2,6-difluoro-3-methyl-benzoic acid,acmc-1af95,3-carboxy-2,4-difluorotoluene,benzoic acid,2,6-difluoro-3-methyl,2,6-difluoro-3-methylbenzoic acid 1g
2774142
2,6-difluoro-3-methylbenzoic acid
CC1=C(C(=C(C=C1)F)C(=O)O)F
Melting Point 138°C to 142°C
Quantity 1g
Formula Weight 172.13
Percent Purity ≥97%
Chemical Name or Material 2,6-Difluoro-3-methylbenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning